Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRPM5
Entrez Gene ID
29850
Gene Name
transient receptor potential cation channel subfamily M member 5
Gene Aliases
LTRPC5, MTR1
Chromosome Location
Chr.11: 2403962 - 2423424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014555.3 NP_055370.1 5 - 6 725 121
XM_017017628.1 XP_016873117.1 6 - 7 812 121
GenBank mRNA AF177473.1 - 5 - 6 725 121
AJ270996.1 - 5 - 6 698 121
BC093787.1 - 5 - 6 722 121
BC093789.1 - 5 - 6 722 121
BC143351.1 - 5 - 6 722 121
BC143353.1 - 5 - 6 722 121

View Full Product Details