The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317960.1 | NP_001304889.1 | 18 - 19 | 2049 | 100 |
NM_012255.4 | NP_036387.2 | 18 - 19 | 1889 | 100 | |
XM_017027723.1 | XP_016883212.1 | 18 - 19 | 1842 | 100 | |
GenBank mRNA | AF064257.1 | - | 18 - 19 | 1842 | 100 |
AF152169.1 | - | 18 - 19 | 1792 | 100 | |
AK000084.1 | - | 7 - 8 | 704 | 100 | |
AK001573.1 | - | 18 - 19 | 1859 | 100 | |
AK302846.1 | - | 18 - 19 | 1753 | 100 | |
AK303312.1 | - | 17 - 18 | 1700 | 100 | |
AK314602.1 | - | 18 - 19 | 1828 | 100 | |
AL136841.1 | - | 18 - 19 | 1815 | 100 | |
AY382900.1 | - | 17 - 18 | 1704 | 100 | |
BC006417.1 | - | 6 - 7 | 664 | 100 |