The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005438.4 | NP_005429.1 | 2 - 3 | 486 | 51 |
GenBank mRNA | AK222902.1 | - | 2 - 3 | 343 | 51 |
AK313718.1 | - | 2 - 3 | 379 | 51 | |
BC016648.1 | - | 2 - 3 | 344 | 51 | |
BQ937004.1 | - | 2 - 3 | 486 | 51 | |
CR542257.1 | - | 2 - 3 | 299 | 51 | |
CR542278.1 | - | 2 - 3 | 299 | 51 | |
KU178407.1 | - | 2 - 3 | 299 | 51 | |
X16707.1 | - | 2 - 3 | 333 | 51 |