The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014763.3 | NP_055578.2 | 3 - 4 | 357 | 150 |
GenBank mRNA | AK291250.1 | - | 3 - 4 | 343 | 150 |
AK295578.1 | - | 3 - 4 | 329 | 150 | |
BC030144.2 | - | 3 - 4 | 342 | 150 | |
BI551514.1 | - | 3 - 4 | 362 | 150 | |
BQ432684.1 | - | 3 - 4 | 360 | 150 | |
D14660.1 | - | 3 - 4 | 330 | 150 |