The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001442.2 | NP_001433.1 | 1 - 2 | 142 | 105 |
GenBank mRNA | BC003672.1 | - | 1 - 2 | 137 | 105 |
BQ880795.1 | - | 1 - 2 | 74 | 105 | |
BT006809.1 | - | 1 - 2 | 71 | 105 | |
CD000452.1 | - | 1 - 2 | 143 | 105 | |
CR456903.1 | - | 1 - 2 | 71 | 105 | |
FJ224319.1 | - | 1 - 2 | 133 | 105 | |
J02874.1 | - | 1 - 2 | 133 | 105 |