The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001567.1 | NP_001001567.1 | 15 - 16 | 1343 | 101 |
NM_001001568.1 | NP_001001568.1 | 13 - 14 | 1194 | 101 | |
NM_001001569.1 | NP_001001569.1 | 12 - 13 | 1139 | 101 | |
NM_001001570.1 | NP_001001570.1 | 15 - 16 | 1364 | 101 | |
NM_001001571.1 | NP_001001571.1 | 14 - 15 | 1272 | 101 | |
NM_001001572.1 | NP_001001572.1 | 12 - 13 | 1150 | 101 | |
NM_001001573.1 | NP_001001573.1 | 13 - 14 | 1330 | 101 | |
NM_001001574.1 | NP_001001574.1 | 14 - 15 | 1265 | 101 | |
NM_001001575.1 | NP_001001575.1 | 11 - 12 | 1095 | 101 | |
NM_001001576.1 | NP_001001576.1 | 11 - 12 | 1054 | 101 | |
NM_001001577.1 | NP_001001577.1 | 12 - 13 | 1173 | 101 | |
NM_001001578.1 | NP_001001578.1 | 13 - 14 | 1217 | 101 | |
NM_001001579.1 | NP_001001579.1 | 13 - 14 | 1210 | 101 | |
NM_001001580.1 | NP_001001580.1 | 10 - 11 | 1010 | 101 | |
NM_001001581.1 | NP_001001581.1 | 14 - 15 | 1374 | 101 | |
NM_001001582.1 | NP_001001582.1 | 15 - 16 | 1452 | 101 | |
NM_001001583.1 | NP_001001583.1 | 15 - 16 | 1445 | 101 | |
NM_001001584.2 | NP_001001584.1 | 14 - 15 | 1273 | 101 | |
NM_001001585.1 | NP_001001585.1 | 14 - 15 | 1397 | 101 | |
NM_001315533.1 | NP_001302462.1 | 16 - 17 | 1419 | 101 | |
NM_002606.2 | NP_002597.1 | 16 - 17 | 1523 | 101 | |
XM_011529598.2 | XP_011527900.1 | 16 - 17 | 1445 | 101 | |
XM_017028366.1 | XP_016883855.1 | 14 - 15 | 1261 | 101 | |
GenBank mRNA | AF048837.1 | - | 16 - 17 | 1520 | 101 |
AF067223.1 | - | 16 - 17 | 1472 | 101 | |
AF067224.1 | - | 15 - 16 | 1291 | 101 | |
AF067225.1 | - | 13 - 14 | 1133 | 101 | |
AF067226.1 | - | 12 - 13 | 1078 | 101 | |
AK001165.1 | - | 13 - 14 | 1125 | 101 | |
AK001307.1 | - | 15 - 16 | 1314 | 101 | |
AK127770.1 | - | 6 - 7 | 2874 | 101 | |
AK294112.1 | - | 13 - 14 | 1160 | 101 | |
AK314679.1 | - | 16 - 17 | 1496 | 101 | |
AY196299.1 | - | 15 - 16 | 1337 | 101 | |
AY196300.1 | - | 14 - 15 | 1245 | 101 | |
AY196301.1 | - | 12 - 13 | 1123 | 101 | |
AY196302.1 | - | 13 - 14 | 1303 | 101 | |
AY196303.1 | - | 14 - 15 | 1238 | 101 | |
AY196304.1 | - | 11 - 12 | 1068 | 101 | |
AY196305.1 | - | 11 - 12 | 1027 | 101 | |
AY196306.1 | - | 12 - 13 | 1146 | 101 | |
AY196307.1 | - | 13 - 14 | 1190 | 101 | |
AY196308.1 | - | 13 - 14 | 1183 | 101 | |
AY196309.1 | - | 10 - 11 | 983 | 101 | |
AY196310.1 | - | 14 - 15 | 1347 | 101 | |
AY196311.1 | - | 15 - 16 | 1425 | 101 | |
AY196312.1 | - | 15 - 16 | 1418 | 101 | |
AY196313.1 | - | 14 - 15 | 1245 | 101 | |
AY196314.1 | - | 14 - 15 | 1370 | 101 | |
AY242121.1 | - | 14 - 15 | 1209 | 101 | |
AY701187.1 | - | 16 - 17 | 1416 | 101 | |
BC009047.1 | - | 15 - 16 | 1317 | 101 | |
BT007016.1 | - | 15 - 16 | 1280 | 101 | |
KU178246.1 | - | 15 - 16 | 1280 | 101 | |
KU178247.1 | - | 16 - 17 | 1460 | 101 | |
KU178248.1 | - | 12 - 13 | 1076 | 101 |