The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242314.1 | NP_001229243.1 | 9 - 10 | 1677 | 78 |
NM_001242317.1 | NP_001229246.1 | 7 - 8 | 941 | 78 | |
NM_004721.4 | NP_004712.1 | 8 - 9 | 1792 | 78 | |
XM_011513310.2 | XP_011511612.1 | 8 - 9 | 1950 | 78 | |
XM_017007456.1 | XP_016862945.1 | 9 - 10 | 1688 | 78 | |
XM_017007457.1 | XP_016862946.1 | 7 - 8 | 1390 | 78 | |
XM_017007458.1 | XP_016862947.1 | 9 - 10 | 1578 | 78 | |
XM_017007459.1 | XP_016862948.1 | 8 - 9 | 1517 | 78 | |
GenBank mRNA | AB001872.1 | - | 8 - 9 | 1658 | 78 |
AK296961.1 | - | 8 - 9 | 1720 | 78 | |
AK297646.1 | - | 7 - 8 | 929 | 78 | |
AK302951.1 | - | 8 - 9 | 1154 | 78 | |
AK312714.1 | - | 9 - 10 | 1642 | 78 | |
BC111726.1 | - | 6 - 7 | 1222 | 78 |