The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164271.1 | NP_001157743.1 | 13 - 14 | 3216 | 95 |
NM_001316668.1 | NP_001303597.1 | 13 - 14 | 3404 | 95 | |
NM_006094.4 | NP_006085.2 | 13 - 14 | 3474 | 95 | |
NM_182643.2 | NP_872584.2 | 17 - 18 | 4909 | 95 | |
XM_005273374.1 | XP_005273431.1 | 17 - 18 | 4790 | 95 | |
XM_017012951.1 | XP_016868440.1 | 17 - 18 | 4660 | 95 | |
XM_017012952.1 | XP_016868441.1 | 13 - 14 | 3207 | 95 | |
GenBank mRNA | AB051510.1 | - | 17 - 18 | 4795 | 95 |
AF026219.1 | - | 13 - 14 | 3427 | 95 | |
AF035119.1 | - | 13 - 14 | 3478 | 95 | |
AK025544.1 | - | 13 - 14 | 3403 | 95 | |
AK025733.1 | - | 8 - 9 | 1650 | 95 | |
AK289734.1 | - | 13 - 14 | 3397 | 95 | |
AK291682.1 | - | 17 - 18 | 4750 | 95 | |
AK299049.1 | - | 13 - 14 | 3216 | 95 | |
BC054511.1 | - | 17 - 18 | 4664 | 95 |