The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007262.4 | NP_009193.2 | 1 - 2 | 140 | 74 |
GenBank mRNA | BC008188.2 | - | 1 - 2 | 61 | 74 |