Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP13-4
Entrez Gene ID
284827
Gene Name
keratin associated protein 13-4
Gene Aliases
KAP13.4
Chromosome Location
Chr.21: 30430276 - 30430758 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181600.1 NP_853631.1 1 - 1 246 100
GenBank mRNA AB096941.1 - 1 - 1 268 100
BC131691.1 - 1 - 1 250 100

View Full Product Details