The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005694.1 | NP_005685.1 | 1 - 1 | 141 | 76 |
GenBank mRNA | AK312143.1 | - | 1 - 1 | 122 | 76 |
BC010933.1 | - | 1 - 1 | 133 | 76 | |
BC105280.1 | - | 1 - 1 | 131 | 76 | |
BC108317.1 | - | 1 - 1 | 148 | 76 | |
L77701.1 | - | 1 - 1 | 141 | 76 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK377174.1 | - | 1 - 1 | 138 | 76 |
AK377176.1 | - | 1 - 1 | 138 | 76 | |
AK377177.1 | - | 1 - 1 | 138 | 76 | |
AK377178.1 | - | 1 - 1 | 138 | 76 | |
AK377179.1 | - | 1 - 1 | 138 | 76 | |
AK377180.1 | - | 1 - 1 | 138 | 76 | |
AK377181.1 | - | 1 - 1 | 138 | 76 | |
AK377182.1 | - | 1 - 1 | 138 | 76 | |
AK377183.1 | - | 1 - 1 | 138 | 76 | |
AK377184.1 | - | 1 - 1 | 138 | 76 |