Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93789932-93790191 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGI1
Entrez Gene ID
9211
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
UniGene
Hs.533670
Gene Chromosome Location
Chr.10: 93757770-93798174 on Build GRCh38
Transcripts
NM_001308275.1 NM_001308276.1 NM_005097.3 NR_131777.1 XM_017016911.1 XM_017016912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978928 c.441A>G p.A147A Chr.10: 93790108-93790108 on Build GRCh38
4697843 c.474T>G p.I158M Chr.10: 93790141-93790141 on Build GRCh38
6130814 c.498A>T p.T166T Chr.10: 93790165-93790165 on Build GRCh38
921626 c.446A>T p.N149I Chr.10: 93790113-93790113 on Build GRCh38
3868087 c.477C>T p.F159F Chr.10: 93790144-93790144 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550004246 Chr.10: 93790018-93790018 on Build GRCh38
rs989516227 Chr.10: 93789968-93789968 on Build GRCh38
rs1052902687 Chr.10: 93790076-93790076 on Build GRCh38
rs769981729 Chr.10: 93790151-93790151 on Build GRCh38
rs139908142 Chr.10: 93790153-93790153 on Build GRCh38
rs921807613 Chr.10: 93789968-93789968 on Build GRCh38
rs912034339 Chr.10: 93789972-93789972 on Build GRCh38
rs866156051 Chr.10: 93790121-93790121 on Build GRCh38
rs371933363 Chr.10: 93790079-93790080 on Build GRCh38
rs756891306 Chr.10: 93789992-93789992 on Build GRCh38
rs953220449 Chr.10: 93789982-93789982 on Build GRCh38
rs781464424 Chr.10: 93790077-93790077 on Build GRCh38
rs935394514 Chr.10: 93790037-93790037 on Build GRCh38
rs1014154783 Chr.10: 93790150-93790150 on Build GRCh38
rs946699377 Chr.10: 93790017-93790017 on Build GRCh38
rs748538032 Chr.10: 93790105-93790105 on Build GRCh38
rs201753712 Chr.10: 93790096-93790096 on Build GRCh38
rs780742254 Chr.10: 93790078-93790079 on Build GRCh38
rs185545799 Chr.10: 93790023-93790023 on Build GRCh38
rs921954988 Chr.10: 93790016-93790016 on Build GRCh38
rs565427077 Chr.10: 93790065-93790066 on Build GRCh38
rs745562947 Chr.10: 93790073-93790073 on Build GRCh38
rs371135873 Chr.10: 93790158-93790158 on Build GRCh38
rs920223409 Chr.10: 93790099-93790099 on Build GRCh38
rs386372133 Chr.10: 93790080-93790081 on Build GRCh38
rs547511256 Chr.10: 93790080-93790080 on Build GRCh38
rs974970902 Chr.10: 93789998-93789998 on Build GRCh38
rs895636677 Chr.10: 93790079-93790079 on Build GRCh38
rs529962843 Chr.10: 93789971-93789971 on Build GRCh38
rs756786865 Chr.10: 93790050-93790050 on Build GRCh38
rs75676567 Chr.10: 93789976-93789976 on Build GRCh38
rs386372134 Chr.10: 93790093-93790094 on Build GRCh38
rs200725559 Chr.10: 93789967-93789968 on Build GRCh38
rs61662431 Chr.10: 93790094-93790095 on Build GRCh38
rs201822203 Chr.10: 93790075-93790075 on Build GRCh38
rs778358284 Chr.10: 93790056-93790056 on Build GRCh38
rs761154452 Chr.10: 93790092-93790093 on Build GRCh38

View Full Product Details