The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198836.1 | NP_001185765.1 | 3 - 3 | 2802 | 77 |
NM_001198837.1 | NP_001185766.1 | 2 - 2 | 2691 | 77 | |
NM_006328.3 | NP_006319.1 | 3 - 3 | 4156 | 77 | |
GenBank mRNA | BC007641.1 | - | 1 - 1 | 751 | 77 |
BC024260.1 | - | 1 - 1 | 751 | 77 | |
CR749306.1 | - | 1 - 1 | 2364 | 77 |