Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
QRSL1
Entrez Gene ID
55278
Gene Name
glutaminyl-tRNA synthase (glutamine-hydrolyzing)-like 1
Gene Aliases
GatA
Chromosome Location
Chr.6: 106629566 - 106668417 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018292.4 NP_060762.3 3 - 4 388 94
XM_011535924.2 XP_011534226.1 4 - 5 482 94
GenBank mRNA AK001851.1 - 3 - 4 370 94
AK022251.1 - 3 - 4 340 94
AK023509.1 - 3 - 4 322 94
AK225262.1 - 3 - 4 376 94
AK225443.1 - 3 - 4 373 94
AK314694.1 - 3 - 4 373 94
AL136679.1 - 3 - 4 376 94
BC006084.1 - 3 - 4 348 94
BC014389.2 - 3 - 4 373 94
DA819470.1 - 3 - 4 388 94

View Full Product Details