The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015488.4 | NP_056303.3 | 8 - 9 | 1014 | 83 |
NM_022572.4 | NP_072094.1 | 7 - 8 | 887 | 83 | |
GenBank mRNA | AB033010.1 | - | 8 - 9 | 855 | 83 |
AK289867.1 | - | 8 - 9 | 922 | 83 | |
AK309308.1 | - | - | 717 | 83 | |
AL137675.1 | - | 6 - 7 | 1513 | 83 | |
AY358753.1 | - | 7 - 8 | 807 | 83 | |
BC002937.2 | - | 7 - 8 | 798 | 83 | |
BC036457.2 | - | 8 - 9 | 916 | 83 |