Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCANP1
Entrez Gene ID
140947
Gene Name
dendritic cell associated nuclear protein
Gene Aliases
C5orf20, DCNP1
Chromosome Location
Chr.5: 135444214 - 135447348 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130848.2 NP_570900.1 1 - 1 3062 76
GenBank mRNA AB074498.1 - 1 - 1 3062 76

View Full Product Details