Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 123060955-123061210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA8
Entrez Gene ID
3312
Gene Name
heat shock protein family A (Hsp70) member 8
Gene Aliases
HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
UniGene
Hs.180414
Gene Chromosome Location
Chr.11: 123057492-123062366 on Build GRCh38
Transcripts
NM_006597.5 NM_153201.3 XM_011542798.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1704728 c.183G>A p.M61I Chr.11: 123061142-123061142 on Build GRCh38
1297759 c.150G>A p.L50L Chr.11: 123061175-123061175 on Build GRCh38
5838041 c.191C>T p.T64I Chr.11: 123061134-123061134 on Build GRCh38
6232515 c.189C>T p.P63P Chr.11: 123061136-123061136 on Build GRCh38
4595947 c.175G>T p.V59F Chr.11: 123061150-123061150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775162274 Chr.11: 123061190-123061190 on Build GRCh38
rs373173372 Chr.11: 123061100-123061100 on Build GRCh38
rs746177143 Chr.11: 123060990-123060990 on Build GRCh38
rs550092655 Chr.11: 123061078-123061078 on Build GRCh38
rs531573882 Chr.11: 123061036-123061036 on Build GRCh38
rs1044324018 Chr.11: 123061037-123061037 on Build GRCh38
rs776800706 Chr.11: 123061043-123061043 on Build GRCh38
rs749956024 Chr.11: 123061111-123061111 on Build GRCh38
rs756843449 Chr.11: 123061097-123061097 on Build GRCh38
rs934474485 Chr.11: 123061013-123061013 on Build GRCh38
rs910169755 Chr.11: 123061005-123061005 on Build GRCh38
rs762594578 Chr.11: 123061044-123061045 on Build GRCh38
rs893047876 Chr.11: 123061022-123061022 on Build GRCh38
rs770830844 Chr.11: 123061079-123061079 on Build GRCh38
rs770980780 Chr.11: 123061186-123061186 on Build GRCh38
rs758689105 Chr.11: 123060998-123060998 on Build GRCh38
rs1002888953 Chr.11: 123061053-123061053 on Build GRCh38
rs199749364 Chr.11: 123061082-123061082 on Build GRCh38
rs747430342 Chr.11: 123061069-123061069 on Build GRCh38
rs768851917 Chr.11: 123061071-123061071 on Build GRCh38
rs560638813 Chr.11: 123061000-123061000 on Build GRCh38
rs763763404 Chr.11: 123061095-123061095 on Build GRCh38
rs369634495 Chr.11: 123061096-123061096 on Build GRCh38
rs747449685 Chr.11: 123061115-123061115 on Build GRCh38
rs183946363 Chr.11: 123061009-123061009 on Build GRCh38
rs1035754137 Chr.11: 123061056-123061056 on Build GRCh38
rs779160391 Chr.11: 123060981-123060981 on Build GRCh38
rs749193179 Chr.11: 123061064-123061064 on Build GRCh38
rs113256342 Chr.11: 123061019-123061020 on Build GRCh38
rs376156706 Chr.11: 123061172-123061172 on Build GRCh38
rs984469760 Chr.11: 123060997-123060997 on Build GRCh38
rs1040069935 Chr.11: 123061008-123061008 on Build GRCh38
rs376044760 Chr.11: 123061083-123061083 on Build GRCh38
rs897272450 Chr.11: 123061059-123061059 on Build GRCh38
rs757940184 Chr.11: 123061112-123061112 on Build GRCh38
rs769131425 Chr.11: 123061130-123061130 on Build GRCh38
rs767193605 Chr.11: 123061163-123061163 on Build GRCh38
rs564184148 Chr.11: 123061021-123061021 on Build GRCh38
rs762957333 Chr.11: 123061081-123061081 on Build GRCh38
rs75167610 Chr.11: 123061001-123061001 on Build GRCh38
rs781396927 Chr.11: 123061160-123061160 on Build GRCh38
rs760570450 Chr.11: 123061086-123061086 on Build GRCh38
rs763047269 Chr.11: 123061180-123061180 on Build GRCh38
rs773294706 Chr.11: 123061080-123061080 on Build GRCh38
rs751780490 Chr.11: 123061046-123061047 on Build GRCh38
rs994330113 Chr.11: 123061073-123061073 on Build GRCh38
rs776953199 Chr.11: 123061075-123061075 on Build GRCh38
rs1018540488 Chr.11: 123061089-123061089 on Build GRCh38

View Full Product Details