The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281727.1 | NP_001268656.1 | 4 - 4 | 474 | 123 |
NM_001281728.1 | NP_001268657.1 | 5 - 5 | 553 | 123 | |
NM_001281729.1 | NP_001268658.1 | 5 - 5 | 861 | 123 | |
NM_014183.3 | NP_054902.1 | 4 - 4 | 478 | 123 | |
NM_177953.2 | NP_808852.1 | 5 - 5 | 557 | 123 | |
NR_104032.1 | 5 - 5 | 560 | 123 | ||
GenBank mRNA | AF161511.1 | - | 4 - 4 | 449 | 123 |
AF165516.1 | - | 4 - 4 | 431 | 123 | |
AK026864.1 | - | 3 - 3 | 414 | 123 | |
BC002481.1 | - | 4 - 4 | 456 | 123 | |
BG110161.1 | - | 4 - 4 | 463 | 123 | |
BG419592.1 | - | 5 - 5 | 529 | 123 | |
BM791453.1 | - | 5 - 5 | 539 | 123 | |
BU163605.1 | - | - | 497 | 123 | |
CB149456.1 | - | 4 - 4 | 474 | 123 |