The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003526.2 | NP_003517.2 | 1 - 1 | 372 | 232 |
GenBank mRNA | BC106899.2 | - | 1 - 1 | 371 | 232 |