The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000018.3 | NP_000009.1 | 13 - 14 | 1487 | 129 |
NM_001033859.2 | NP_001029031.1 | 12 - 13 | 1421 | 129 | |
NM_001270447.1 | NP_001257376.1 | 14 - 15 | 1423 | 129 | |
NM_001270448.1 | NP_001257377.1 | 12 - 13 | 1562 | 129 | |
XM_006721516.2 | XP_006721579.2 | 13 - 14 | 1440 | 129 | |
XM_011523829.1 | XP_011522131.1 | 13 - 14 | 1440 | 129 | |
XM_011523830.1 | XP_011522132.1 | 13 - 14 | 1440 | 129 | |
GenBank mRNA | AK056083.1 | - | 12 - 13 | 1455 | 129 |
AK056956.1 | - | 8 - 9 | 833 | 129 | |
AK097243.1 | - | 10 - 11 | 2169 | 129 | |
AK222518.1 | - | 13 - 14 | 1369 | 129 | |
AK293549.1 | - | 14 - 15 | 1423 | 129 | |
BC000399.2 | - | 13 - 14 | 1425 | 129 | |
BC012912.1 | - | 13 - 14 | 1409 | 129 | |
BC020218.1 | - | 12 - 13 | 1329 | 129 | |
D43682.1 | - | 13 - 14 | 1366 | 129 | |
X86556.1 | - | 13 - 14 | 1418 | 129 |