Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SFI1
- Entrez Gene ID
-
9814
- Gene Name
- SFI1 centrin binding protein
- Gene Aliases
-
PISD, PPP1R139, hSfi1p
- Chromosome Location
-
Chr.22: 31496139 - 31618554 on Build GRCh38
View Full Product Details