The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001031679.2 | NP_001026849.1 | 5 - 6 | 565 | 107 |
NM_001193460.1 | NP_001180389.1 | 6 - 7 | 729 | 107 | |
NM_001193461.1 | NP_001180390.1 | 5 - 6 | 427 | 107 | |
NM_198080.3 | NP_932346.1 | 4 - 5 | 438 | 107 | |
GenBank mRNA | AK293084.1 | - | 5 - 6 | 535 | 107 |
AK299065.1 | - | 4 - 5 | 377 | 107 | |
AK316365.1 | - | 4 - 5 | 409 | 107 | |
AL833622.1 | - | 5 - 6 | 483 | 107 | |
AY358229.1 | - | 4 - 5 | 373 | 107 | |
BC040053.1 | - | 6 - 7 | 630 | 107 | |
BX500552.1 | - | 5 - 6 | 429 | 107 | |
BX648776.1 | - | 5 - 6 | 484 | 107 | |
DC357690.1 | - | 4 - 5 | 438 | 107 |