The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006.4 | NP_000997.1 | 6 - 6 | 830 | 155 |
GenBank mRNA | BC004981.1 | - | 6 - 6 | 762 | 155 |