The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001082575.2 | NP_001076044.1 | 5 - 6 | 739 | 78 |
XM_011524359.1 | XP_011522661.1 | 5 - 6 | 821 | 78 | |
XM_011524360.1 | XP_011522662.1 | 6 - 7 | 1010 | 78 | |
XM_011524363.2 | XP_011522665.1 | 5 - 6 | 821 | 78 | |
XM_011524365.2 | XP_011522667.1 | 5 - 6 | 821 | 78 | |
XM_011524367.2 | XP_011522669.1 | 5 - 6 | 821 | 78 | |
XM_017024208.1 | XP_016879697.1 | 4 - 5 | 781 | 78 | |
XM_017024209.1 | XP_016879698.1 | 5 - 6 | 821 | 78 | |
XM_017024210.1 | XP_016879699.1 | 5 - 6 | 821 | 78 | |
XM_017024211.1 | XP_016879700.1 | 5 - 6 | 821 | 78 | |
GenBank mRNA | AK293617.1 | - | 5 - 6 | 739 | 78 |
AK293905.1 | - | 5 - 6 | 602 | 78 | |
BX452143.2 | - | 1 - 2 | 261 | 78 | |
DN990270.1 | - | 4 - 5 | 438 | 78 |