The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164239.1 | NP_001157711.1 | 14 - 15 | 2200 | 70 |
NM_003587.4 | NP_003578.2 | 14 - 15 | 2521 | 70 | |
XM_011514938.2 | XP_011513240.1 | 11 - 12 | 2314 | 70 | |
XM_011514939.2 | XP_011513241.1 | 11 - 12 | 1788 | 70 | |
XM_011514940.2 | XP_011513242.1 | 14 - 15 | 2729 | 70 | |
XM_011514941.2 | XP_011513243.1 | 6 - 7 | 813 | 70 | |
GenBank mRNA | AB001601.1 | - | 14 - 15 | 2448 | 70 |
AF119860.1 | - | 6 - 7 | 993 | 70 | |
AK302721.1 | - | 14 - 15 | 2194 | 70 | |
AK312264.1 | - | 14 - 15 | 2460 | 70 | |
BC008825.2 | - | 14 - 15 | 2472 | 70 | |
BC009392.2 | - | 14 - 15 | 2472 | 70 |