The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005069.4 | NP_005060.1 | 6 - 7 | 1356 | 68 |
NM_009586.3 | NP_033664.2 | 6 - 7 | 1356 | 68 | |
XM_011529694.1 | XP_011527996.1 | 5 - 6 | 1523 | 68 | |
XM_017028442.1 | XP_016883931.1 | 6 - 7 | 1372 | 68 | |
GenBank mRNA | AJ001858.1 | - | 5 - 6 | 554 | 68 |
BC110444.2 | - | 6 - 7 | 768 | 68 | |
U80456.1 | - | 6 - 7 | 834 | 68 | |
U80457.1 | - | 6 - 7 | 834 | 68 |