Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- THRB
- Entrez Gene ID
-
7068
- Gene Name
- thyroid hormone receptor beta
- Gene Aliases
-
C-ERBA-2, C-ERBA-BETA, ERBA2, GRTH, NR1A2, PRTH, THR1, THRB1, THRB2
- Chromosome Location
-
Chr.3: 24117153 - 24495281 on Build GRCh38
View Full Product Details