Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLM1
Entrez Gene ID
51280
Gene Name
golgi membrane protein 1
Gene Aliases
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3
Chromosome Location
Chr.9: 86026141 - 86100201 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016548.3 NP_057632.2 9 - 10 1326 115
NM_177937.2 NP_808800.1 9 - 10 1318 115
GenBank mRNA AF236056.1 - 9 - 10 1267 115
AK027717.1 - 9 - 10 1289 115
AK075542.1 - 9 - 10 1260 115
AK075557.1 - 9 - 10 1230 115
AK092943.1 - 10 - 11 1206 115
AY358593.1 - 9 - 10 1231 115
BC001740.1 - 9 - 10 1241 115
CR457201.1 - 8 - 9 1117 115
EU794616.1 - 9 - 10 1285 115

View Full Product Details