The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000233.3 | NP_000224.2 | 3 - 4 | 374 | 84 |
XM_017004089.1 | XP_016859578.1 | 1 - 2 | 2062 | 84 | |
GenBank mRNA | M63108.1 | - | 3 - 4 | 312 | 84 |
M73746.1 | - | 3 - 4 | 336 | 84 | |
S57793.1 | - | 3 - 4 | 304 | 84 |