The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244710.1 | NP_001231639.1 | 3 - 4 | 406 | 127 |
NM_002056.3 | NP_002047.2 | 3 - 4 | 406 | 127 | |
XM_017003801.1 | XP_016859290.1 | 3 - 4 | 370 | 127 | |
XM_017003802.1 | XP_016859291.1 | 3 - 4 | 362 | 127 | |
GenBank mRNA | BC045641.1 | - | 3 - 4 | 402 | 127 |
BG723175.1 | - | 3 - 4 | 410 | 129 | |
BI560595.1 | - | 3 - 4 | 414 | 127 | |
DB133199.1 | - | 2 - 3 | 202 | 127 |