The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025930.3 | NP_001021100.3 | 6 - 7 | 1009 | 105 |
NR_037162.1 | 6 - 7 | 1631 | 105 | ||
GenBank mRNA | AK023960.1 | - | 6 - 7 | 626 | 105 |
AK297289.1 | - | 3 - 4 | 353 | 105 | |
AK315140.1 | - | 4 - 5 | 399 | 105 | |
AL833939.1 | - | 2 - 3 | 1436 | 105 | |
BP357921.1 | - | 4 - 5 | 385 | 105 |