The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128177.1 | NP_001121649.1 | 4 - 5 | 444 | 101 |
XM_011534052.2 | XP_011532354.1 | 4 - 5 | 330 | 101 |