The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177599.1 | NP_001171070.1 | 7 - 8 | 791 | 79 |
NM_003848.3 | NP_003839.2 | 7 - 8 | 791 | 79 | |
XM_017007415.1 | XP_016862904.1 | 7 - 8 | 787 | 79 | |
XM_017007416.1 | XP_016862905.1 | 7 - 8 | 787 | 79 | |
XM_017007417.1 | XP_016862906.1 | 7 - 8 | 787 | 79 | |
XM_017007418.1 | XP_016862907.1 | 7 - 8 | 787 | 79 | |
XM_017007419.1 | XP_016862908.1 | 7 - 8 | 787 | 79 | |
XM_017007420.1 | XP_016862909.1 | 7 - 8 | 787 | 79 | |
GenBank mRNA | AF058954.1 | - | 6 - 7 | 679 | 79 |
AF131748.1 | - | 1 - 2 | 64 | 79 | |
AK294598.1 | - | 7 - 8 | 784 | 79 | |
AK310527.1 | - | 7 - 8 | 787 | 79 | |
BC007716.2 | - | 7 - 8 | 754 | 79 | |
BC019868.1 | - | 4 - 5 | 430 | 79 | |
BC035149.1 | - | 6 - 7 | 553 | 79 | |
BC047024.1 | - | 8 - 9 | 761 | 79 | |
BC068602.1 | - | 7 - 8 | 792 | 79 |