Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VGLL4
Entrez Gene ID
9686
Gene Name
vestigial like family member 4
Gene Aliases
VGL-4
Chromosome Location
Chr.3: 11556067 - 11720768 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284391.1 NP_001271320.1 1 - 2 260 111
GenBank mRNA AK300344.1 - 1 - 2 260 111

View Full Product Details