The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005260257.1 | XP_005260314.1 | 1 - 2 | 91 | 69 |
XM_017027603.1 | XP_016883092.1 | 1 - 2 | 91 | 69 | |
GenBank mRNA | HY029018.1 | - | 1 - 2 | 97 | 69 |