Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD1
Entrez Gene ID
5707
Gene Name
proteasome 26S subunit, non-ATPase 1
Gene Aliases
P112, Rpn2, S1
Chromosome Location
Chr.2: 231056864 - 231172827 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191037.1 NP_001177966.1 18 - 19 2274 109
NM_002807.3 NP_002798.2 18 - 19 2274 109
NR_034059.1 17 - 18 2104 109
GenBank mRNA AK307415.1 - 17 - 18 2101 109
AK307513.1 - 16 - 17 1976 109
AK312528.1 - 18 - 19 2231 109
BC001053.2 - 18 - 19 2231 109
BC014013.2 - 18 - 19 2243 109
BC020223.1 - 18 - 19 2221 109
BC032445.1 - 18 - 19 2246 109
BC039845.1 - 18 - 19 2197 109
BC047897.1 - 18 - 19 2196 109
BC064398.1 - 18 - 19 2245 109
BC073833.1 - 18 - 19 2198 109
BC094720.1 - 18 - 19 2260 109
BC112344.1 - 18 - 19 2235 109
BC114434.1 - 18 - 19 2235 109
D44466.1 - 18 - 19 2192 109

View Full Product Details