Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNE1
Entrez Gene ID
3753
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 1
Gene Aliases
ISK, JLNS, JLNS2, LQT2/5, LQT5, MinK
Chromosome Location
Chr.21: 34446688 - 34512275 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127669.3 NP_001121141.1 1 - 2 71 61
GenBank mRNA BC036452.1 - 1 - 2 74 61
EU008569.1 - 1 - 2 62 61

View Full Product Details