The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 26 - 27 | 2803 | 120 |
NM_001282899.1 | NP_001269828.1 | 26 - 27 | 2674 | 120 | |
NM_001282900.1 | NP_001269829.1 | 25 - 26 | 2620 | 120 | |
NM_001282901.1 | NP_001269830.1 | 23 - 24 | 2408 | 120 | |
NM_007023.3 | NP_008954.2 | 29 - 30 | 3044 | 120 | |
XM_005246246.2 | XP_005246303.1 | 28 - 29 | 2857 | 120 | |
XM_006712205.3 | XP_006712268.1 | 28 - 29 | 2862 | 120 | |
XM_011510505.2 | XP_011508807.1 | 28 - 29 | 2891 | 120 | |
XM_017003197.1 | XP_016858686.1 | 25 - 26 | 2560 | 120 | |
GenBank mRNA | AB027471.1 | - | 29 - 30 | 2914 | 120 |
AB075513.1 | - | 13 - 14 | 1259 | 120 | |
AB209681.1 | - | 29 - 30 | 5098 | 120 | |
AK294433.1 | - | 23 - 24 | 2332 | 120 | |
AK294445.1 | - | 26 - 27 | 2523 | 120 | |
AK294914.1 | - | 23 - 24 | 2361 | 120 | |
AK296340.1 | - | 25 - 26 | 2615 | 120 | |
AK304278.1 | - | 26 - 27 | 2563 | 120 | |
AK313084.1 | - | 29 - 30 | 2913 | 120 | |
AK316014.1 | - | 26 - 27 | 2638 | 120 | |
AK316072.1 | - | 26 - 27 | 2570 | 120 | |
BC024004.2 | - | 29 - 30 | 2883 | 120 | |
BC040183.2 | - | 26 - 27 | 2804 | 120 | |
KJ534924.1 | - | 29 - 30 | 2821 | 120 | |
KJ534925.1 | - | 28 - 29 | 2803 | 120 | |
KJ534978.1 | - | 29 - 30 | 2857 | 120 | |
U78516.1 | - | 29 - 30 | 2965 | 120 |