The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 8 - 9 | 1038 | 128 |
NM_001282899.1 | NP_001269828.1 | 8 - 9 | 909 | 128 | |
NM_001282900.1 | NP_001269829.1 | 7 - 8 | 855 | 128 | |
NM_001282901.1 | NP_001269830.1 | 5 - 6 | 643 | 128 | |
NM_007023.3 | NP_008954.2 | 11 - 12 | 1279 | 128 | |
XM_005246246.2 | XP_005246303.1 | 10 - 11 | 1092 | 128 | |
XM_006712205.3 | XP_006712268.1 | 10 - 11 | 1097 | 128 | |
XM_011510505.2 | XP_011508807.1 | 10 - 11 | 1126 | 128 | |
XM_017003196.1 | XP_016858685.1 | 11 - 12 | 1146 | 128 | |
XM_017003197.1 | XP_016858686.1 | 7 - 8 | 795 | 128 | |
GenBank mRNA | AB027471.1 | - | 11 - 12 | 1149 | 128 |
AB209681.1 | - | 11 - 12 | 1208 | 128 | |
AK294433.1 | - | 5 - 6 | 567 | 128 | |
AK294445.1 | - | 8 - 9 | 758 | 128 | |
AK294914.1 | - | 5 - 6 | 596 | 128 | |
AK294997.1 | - | 11 - 12 | 1244 | 128 | |
AK295002.1 | - | 7 - 8 | 790 | 128 | |
AK296340.1 | - | 7 - 8 | 850 | 128 | |
AK302732.1 | - | 6 - 7 | 634 | 128 | |
AK304278.1 | - | 8 - 9 | 798 | 128 | |
AK311575.1 | - | 6 - 7 | 697 | 128 | |
AK313084.1 | - | 11 - 12 | 1148 | 128 | |
AK316072.1 | - | 8 - 9 | 805 | 128 | |
BC024004.2 | - | 11 - 12 | 1118 | 128 | |
BC040183.2 | - | 8 - 9 | 1039 | 128 | |
KJ534924.1 | - | 11 - 12 | 1056 | 128 | |
KJ534925.1 | - | 10 - 11 | 1038 | 128 | |
KJ534978.1 | - | 11 - 12 | 1092 | 128 | |
U78516.1 | - | 11 - 12 | 1200 | 128 |