The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256864.1 | NP_001243793.1 | 5 - 6 | 876 | 85 |
NM_001256865.1 | NP_001243794.1 | 6 - 7 | 824 | 85 | |
NM_014787.3 | NP_055602.1 | 5 - 6 | 711 | 85 | |
GenBank mRNA | AB007942.1 | - | 5 - 6 | 651 | 85 |
BC051722.1 | - | 5 - 6 | 660 | 85 | |
BC109279.2 | - | 5 - 6 | 809 | 85 | |
BC109280.1 | - | 5 - 6 | 812 | 85 | |
BX647203.1 | - | 5 - 6 | 865 | 85 |