Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISPD
Entrez Gene ID
729920
Gene Name
isoprenoid synthase domain containing
Gene Aliases
MDDGA7, MDDGC7, Nip, hCG_1745121
Chromosome Location
Chr.7: 16087527 - 16422562 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101417.3 NP_001094887.1 7 - 8 970 102
NM_001101426.3 NP_001094896.1 8 - 9 1120 102
XM_006715770.3 XP_006715833.1 8 - 9 1290 102
XM_011515500.2 XP_011513802.1 7 - 8 1160 102
XM_011515508.2 XP_011513810.1 8 - 9 1622 102
XM_011515509.2 XP_011513811.1 8 - 9 2471 102
XM_017012575.1 XP_016868064.1 8 - 9 1265 102
XM_017012577.1 XP_016868066.1 7 - 8 1382 102
XM_017012578.1 XP_016868067.1 6 - 7 1797 102
GenBank mRNA DY654823.1 - 5 - 6 373 102

View Full Product Details