Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK12
Entrez Gene ID
51755
Gene Name
cyclin dependent kinase 12
Gene Aliases
CRK7, CRKR, CRKRS
Chromosome Location
Chr.17: 39461486 - 39567560 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016507.3 NP_057591.2 13 - 14 4323 116
XM_005257456.3 XP_005257513.1 13 - 14 4345 116
XM_011524892.2 XP_011523194.1 12 - 13 4270 116
XM_011524893.2 XP_011523195.1 13 - 14 4348 116
XM_011524894.2 XP_011523196.1 13 - 14 4348 116
XM_011524895.2 XP_011523197.1 13 - 14 4348 116
XM_011524896.2 XP_011523198.1 13 - 14 4348 116
XM_011524897.2 XP_011523199.1 13 - 14 4348 116
XM_011524898.2 XP_011523200.1 13 - 14 4348 116
XM_011524899.2 XP_011523201.1 13 - 14 4348 116
XM_011524900.2 XP_011523202.1 13 - 14 4348 116
XM_011524901.2 XP_011523203.1 13 - 14 4348 116
XM_011524902.2 XP_011523204.1 13 - 14 4348 116
XM_011524903.2 XP_011523205.1 13 - 14 4348 116
XM_011524905.2 XP_011523207.1 13 - 14 4348 116
XM_011524906.2 XP_011523208.1 13 - 14 4348 116
XM_017024744.1 XP_016880233.1 13 - 14 4348 116
XM_017024745.1 XP_016880234.1 13 - 14 4348 116
XM_017024746.1 XP_016880235.1 13 - 14 4348 116
XM_017024747.1 XP_016880236.1 13 - 14 4348 116
XM_017024748.1 XP_016880237.1 13 - 14 4348 116
XM_017024749.1 XP_016880238.1 13 - 14 4348 116
GenBank mRNA AF227198.1 - 13 - 14 3795 116

View Full Product Details