Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CDK12
- Entrez Gene ID
-
51755
- Gene Name
- cyclin dependent kinase 12
- Gene Aliases
-
CRK7, CRKR, CRKRS
- Chromosome Location
-
Chr.17: 39461486 - 39567560 on Build GRCh38
View Full Product Details