The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 12 - 13 | 1068 | 97 |
XM_011528487.2 | XP_011526789.1 | 12 - 13 | 1104 | 97 | |
XM_011528488.2 | XP_011526790.1 | 12 - 13 | 1056 | 97 | |
XM_011528489.2 | XP_011526791.1 | 12 - 13 | 1099 | 97 | |
XM_011528490.2 | XP_011526792.1 | 12 - 13 | 1080 | 97 | |
XM_011528493.2 | XP_011526795.1 | 11 - 12 | 1019 | 97 | |
XM_017027586.1 | XP_016883075.1 | 11 - 12 | 1300 | 97 | |
XM_017027587.1 | XP_016883076.1 | 11 - 12 | 1297 | 97 | |
XM_017027588.1 | XP_016883077.1 | 11 - 12 | 1301 | 97 | |
XM_017027589.1 | XP_016883078.1 | 11 - 12 | 1301 | 97 | |
XM_017027590.1 | XP_016883079.1 | 11 - 12 | 993 | 97 | |
XM_017027591.1 | XP_016883080.1 | 4 - 5 | 460 | 97 | |
XM_017027592.1 | XP_016883081.1 | 11 - 12 | 1302 | 97 | |
GenBank mRNA | AK308889.1 | - | 10 - 11 | 1316 | 97 |
BC040566.1 | - | 12 - 13 | 1104 | 97 | |
BC132864.1 | - | 11 - 12 | 937 | 97 | |
BC132870.1 | - | 11 - 12 | 939 | 97 | |
BC146798.1 | - | 12 - 13 | 1056 | 97 | |
BX648091.1 | - | 12 - 13 | 1022 | 97 | |
BX649108.1 | - | 12 - 13 | 1048 | 97 | |
Y08982.1 | - | 10 - 11 | 847 | 97 |