The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 33 - 34 | 3386 | 149 |
XM_011528487.2 | XP_011526789.1 | 33 - 34 | 3422 | 149 | |
XM_011528488.2 | XP_011526790.1 | 33 - 34 | 3374 | 149 | |
XM_011528489.2 | XP_011526791.1 | 33 - 34 | 3417 | 149 | |
XM_011528490.2 | XP_011526792.1 | 33 - 34 | 3398 | 149 | |
XM_011528493.2 | XP_011526795.1 | 32 - 33 | 3337 | 149 | |
XM_017027586.1 | XP_016883075.1 | 32 - 33 | 3618 | 149 | |
XM_017027587.1 | XP_016883076.1 | 32 - 33 | 3615 | 149 | |
XM_017027588.1 | XP_016883077.1 | 32 - 33 | 3619 | 149 | |
XM_017027590.1 | XP_016883079.1 | 32 - 33 | 3311 | 149 | |
XM_017027591.1 | XP_016883080.1 | 24 - 25 | 2717 | 149 | |
GenBank mRNA | BC132864.1 | - | 32 - 33 | 3255 | 149 |
BC132870.1 | - | 32 - 33 | 3257 | 149 | |
BX648091.1 | - | 33 - 34 | 3339 | 149 | |
BX649108.1 | - | 33 - 34 | 3364 | 149 | |
Y08982.1 | - | 31 - 32 | 3165 | 149 |