The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014258.3 | NP_055073.2 | 8 - 9 | 742 | 82 |
XM_011528487.2 | XP_011526789.1 | 8 - 9 | 778 | 82 | |
XM_011528488.2 | XP_011526790.1 | 8 - 9 | 730 | 82 | |
XM_011528489.2 | XP_011526791.1 | 8 - 9 | 773 | 82 | |
XM_011528490.2 | XP_011526792.1 | 8 - 9 | 754 | 82 | |
XM_017027586.1 | XP_016883075.1 | 7 - 8 | 974 | 82 | |
XM_017027587.1 | XP_016883076.1 | 7 - 8 | 971 | 82 | |
XM_017027588.1 | XP_016883077.1 | 7 - 8 | 975 | 82 | |
XM_017027589.1 | XP_016883078.1 | 7 - 8 | 975 | 82 | |
XM_017027592.1 | XP_016883081.1 | 7 - 8 | 976 | 82 | |
GenBank mRNA | AK308889.1 | - | 6 - 7 | 990 | 82 |
BC040566.1 | - | 8 - 9 | 778 | 82 | |
BC132864.1 | - | 7 - 8 | 611 | 82 | |
BC132870.1 | - | 7 - 8 | 613 | 82 | |
BC146798.1 | - | 8 - 9 | 730 | 82 | |
BX648091.1 | - | 8 - 9 | 696 | 82 | |
BX649108.1 | - | 8 - 9 | 722 | 82 | |
Y08982.1 | - | 6 - 7 | 521 | 82 |