The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079864.2 | NP_001073333.1 | 15 - 16 | 2140 | 65 |
NM_001206901.1 | NP_001193830.1 | 15 - 16 | 2143 | 65 | |
NM_001206902.1 | NP_001193831.1 | 14 - 15 | 1952 | 65 | |
NM_006024.6 | NP_006015.4 | 15 - 16 | 2266 | 65 | |
XM_017012754.1 | XP_016868243.1 | 15 - 16 | 2236 | 65 | |
GenBank mRNA | AF090891.1 | - | 15 - 16 | 2058 | 65 |
AF268075.1 | - | 14 - 15 | 1958 | 65 | |
AK000098.1 | - | 15 - 16 | 2179 | 65 | |
AK296720.1 | - | 14 - 15 | 1901 | 65 | |
AK299729.1 | - | 15 - 16 | 2143 | 65 | |
AK301064.1 | - | 15 - 16 | 1929 | 65 | |
AK309757.1 | - | 4 - 5 | 354 | 65 | |
AK314292.1 | - | 15 - 16 | 2236 | 65 | |
AL136586.1 | - | 15 - 16 | 2172 | 65 | |
BC050358.2 | - | 15 - 16 | 2151 | 65 | |
CR457056.1 | - | 14 - 15 | 1958 | 65 | |
CR533556.1 | - | 14 - 15 | 2084 | 65 | |
U33821.2 | - | 15 - 16 | 2041 | 65 |