The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080399.2 | NP_001073868.2 | 11 - 12 | 948 | 79 |
GenBank mRNA | AK297911.1 | - | 12 - 13 | 948 | 79 |