The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080399.2 | NP_001073868.2 | 3 - 4 | 190 | 128 |
GenBank mRNA | AK297911.1 | - | 4 - 5 | 190 | 128 |