The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080399.2 | NP_001073868.2 | 5 - 6 | 430 | 67 |
GenBank mRNA | AK297911.1 | - | 6 - 7 | 430 | 67 |